Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NHX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111512.3 | NP_187288.2 | 14 - 15 | 1684 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |