Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337693.1 | NP_001326712.1 | 20 - 21 | 2316 | 69 | |
NM_001337694.1 | NP_001326713.1 | 19 - 20 | 2184 | 69 | ||
NM_111596.6 | NP_187372.5 | 19 - 20 | 2120 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |