Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATAB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111680.4 | NP_566327.1 | 2 - 3 | 629 | 108 | |
GenBank mRNA | AY072316.1 | - | 2 - 3 | 571 | 108 | |
BT001168.1 | - | 2 - 3 | 553 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |