Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VHA-E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111690.2 | NP_187468.1 | 3 - 4 | 609 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Arabidopsis thaliana Probe spans exons |