Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118552.4 | NP_194150.1 | 12 - 13 | 1840 | 126 | |
NM_202877.1 | NP_974606.1 | 12 - 13 | 1687 | 126 | ||
GenBank mRNA | AB064528.1 | - | 12 - 13 | 1673 | 126 | |
AY039895.1 | - | 12 - 13 | 1681 | 126 | ||
BT004527.1 | - | 12 - 13 | 1633 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |