Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118552.4 | NP_194150.1 | 14 - 15 | 2389 | 81 | |
NM_202877.1 | NP_974606.1 | 14 - 15 | 2236 | 81 | ||
GenBank mRNA | AB064528.1 | - | 14 - 15 | 2222 | 81 | |
AY039895.1 | - | 14 - 15 | 2230 | 81 | ||
BT004527.1 | - | 14 - 15 | 2182 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |