Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341680.1 | NP_001320057.1 | 6 - 7 | 850 | 62 | |
NM_001341681.1 | NP_001329908.1 | 7 - 8 | 735 | 62 | ||
NM_001341682.1 | NP_001329909.1 | 7 - 8 | 507 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |