Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341724.1 | NP_001328533.1 | 5 - 6 | 799 | 84 | |
NM_118655.4 | NP_194253.2 | 5 - 6 | 913 | 84 | ||
NM_179221.2 | NP_849552.1 | 4 - 5 | 1127 | 84 | ||
GenBank mRNA | AY072178.1 | - | 4 - 5 | 914 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |