Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341750.1 | NP_001328550.1 | 1 - 2 | 174 | 125 | |
NM_118688.3 | NP_194286.1 | 2 - 3 | 312 | 125 | ||
GenBank mRNA | AY519609.1 | - | 2 - 3 | 260 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |