Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VATG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118728.4 | NP_194325.1 | 2 - 3 | 381 | 126 | |
GenBank mRNA | AY063859.1 | - | 2 - 3 | 195 | 126 | |
AY091217.1 | - | 2 - 3 | 185 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |