Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIOX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203915.2 | NP_001190844.1 | 3 - 4 | 387 | 142 | |
NM_118759.5 | NP_194356.2 | 3 - 4 | 492 | 142 | ||
GenBank mRNA | AY142501.1 | - | 3 - 4 | 248 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |