Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSN6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118776.3 | NP_567746.1 | 5 - 6 | 859 | 144 | |
GenBank mRNA | AF395064.1 | - | 5 - 6 | 622 | 144 | |
AY087556.1 | - | 5 - 6 | 629 | 144 | ||
BT002345.1 | - | 5 - 6 | 622 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |