Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118793.3 | NP_194389.1 | 1 - 2 | 358 | 70 | |
GenBank mRNA | AF360292.1 | - | 1 - 2 | 268 | 70 | |
AY051048.1 | - | 1 - 2 | 63 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |