Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D6PKL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118795.4 | NP_194391.1 | 2 - 3 | 1101 | 62 | |
GenBank mRNA | AY063923.1 | - | 2 - 3 | 970 | 62 | |
AY091248.1 | - | 1 - 2 | 678 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |