Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118803.5 | NP_567755.1 | 1 - 2 | 338 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |