Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FIM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036649.1 | NP_001031726.1 | 8 - 9 | 1216 | 148 | |
NM_001036650.2 | NP_001031727.1 | 7 - 8 | 1303 | 148 | ||
NM_001341826.1 | NP_001328753.1 | 8 - 9 | 1343 | 148 | ||
NM_001341827.1 | NP_001328754.1 | 8 - 9 | 1327 | 148 | ||
NM_001341828.1 | NP_001328752.1 | 8 - 9 | 1315 | 148 | ||
NM_001341829.1 | NP_001328755.1 | 6 - 7 | 1237 | 148 | ||
NM_118804.5 | NP_194400.1 | 7 - 8 | 1167 | 148 | ||
GenBank mRNA | AK318682.1 | - | 8 - 9 | 1219 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |