Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125593.1 | NP_001119065.1 | 8 - 9 | 1344 | 70 | |
NM_001125594.1 | NP_001119066.1 | 8 - 9 | 1157 | 70 | ||
NM_001125595.2 | NP_001119067.1 | 10 - 11 | 1260 | 70 | ||
NM_001341834.1 | NP_001328604.1 | 8 - 9 | 1522 | 70 | ||
NM_001341835.1 | NP_001328603.1 | 8 - 9 | 1502 | 70 | ||
NM_118838.5 | NP_194434.2 | 10 - 11 | 1311 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |