Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COBL11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118845.3 | NP_567766.1 | 1 - 2 | 638 | 60 | |
GenBank mRNA | AK175703.1 | - | 1 - 2 | 604 | 60 | |
AK176063.1 | - | 1 - 2 | 604 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |