Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RD26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084983.1 | NP_001078452.1 | 2 - 3 | 497 | 124 | |
NM_118875.4 | NP_567773.1 | 2 - 3 | 603 | 124 | ||
NM_179126.1 | NP_849457.1 | 1 - 2 | 403 | 124 | ||
GenBank mRNA | AF083738.1 | - | 2 - 3 | 540 | 124 | |
AF428375.1 | - | 2 - 3 | 547 | 124 | ||
AY057578.1 | - | 2 - 3 | 549 | 124 | ||
AY087772.1 | - | 2 - 3 | 557 | 124 | ||
AY091428.1 | - | 2 - 3 | 446 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |