Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339369.1 | NP_001326576.1 | 1 - 2 | 180 | 90 | |
NM_148863.4 | NP_680116.2 | 4 - 5 | 1536 | 90 | ||
GenBank mRNA | AY664499.1 | - | 4 - 5 | 1183 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |