Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112073.5 | NP_566422.4 | 2 - 3 | 320 | 75 | |
GenBank mRNA | FJ850974.1 | - | 2 - 3 | 170 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |