Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULTR4;2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202942.1 | NP_001189871.1 | 4 - 5 | 532 | 115 | |
NM_001337994.1 | NP_001327651.1 | 3 - 4 | 719 | 115 | ||
NM_112087.3 | NP_187858.1 | 3 - 4 | 794 | 115 | ||
GenBank mRNA | AB052775.1 | - | 3 - 4 | 577 | 115 | |
AK117615.1 | - | 4 - 5 | 538 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |