Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035603.1 | NP_001030680.1 | 7 - 8 | 593 | 61 | |
NM_001337995.1 | NP_001319531.1 | 9 - 10 | 796 | 61 | ||
NM_001337996.1 | NP_001326331.1 | 8 - 9 | 802 | 61 | ||
NM_112088.3 | NP_187859.2 | 8 - 9 | 918 | 61 | ||
GenBank mRNA | BT010931.1 | - | 8 - 9 | 677 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |