Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSP70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112093.3 | NP_187864.1 | 1 - 2 | 405 | 81 | |
GenBank mRNA | AY054183.1 | - | 1 - 2 | 310 | 81 | |
AY054190.1 | - | 1 - 2 | 309 | 81 | ||
AY059885.1 | - | 1 - 2 | 309 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |