Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IAA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084991.1 | NP_001078460.1 | 1 - 2 | 412 | 119 | |
NM_001203930.2 | NP_001190859.1 | 1 - 2 | 484 | 119 | ||
NM_119006.3 | NP_194593.1 | 1 - 2 | 412 | 119 | ||
GenBank mRNA | AF332397.1 | - | 1 - 2 | 259 | 119 | |
AK118377.1 | - | 1 - 2 | 398 | 119 | ||
U18413.1 | - | 1 - 2 | 302 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |