Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119039.3 | NP_194624.1 | 7 - 8 | 967 | 74 | |
GenBank mRNA | AF079484.1 | - | 7 - 8 | 764 | 74 | |
BT025271.1 | - | 7 - 8 | 600 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |