Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119065.3 | NP_194650.1 | 2 - 3 | 517 | 77 | |
NM_202907.2 | NP_974636.1 | 2 - 3 | 529 | 77 | ||
GenBank mRNA | AK228574.1 | - | 2 - 3 | 538 | 77 | |
BT005754.1 | - | 2 - 3 | 517 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |