Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341994.1 | NP_001328455.1 | 2 - 3 | 385 | 75 | |
NM_119158.4 | NP_194741.2 | 3 - 4 | 678 | 75 | ||
GenBank mRNA | DQ446885.1 | - | 3 - 4 | 552 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |