Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNGC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203944.2 | NP_001190873.1 | 3 - 4 | 926 | 121 | |
NM_001342012.1 | NP_001329898.1 | 2 - 3 | 1055 | 121 | ||
NM_001342013.1 | NP_001329900.1 | 3 - 4 | 1043 | 121 | ||
NM_001342014.1 | NP_001329899.1 | 1 - 2 | 642 | 121 | ||
NM_119202.4 | NP_194785.1 | 3 - 4 | 907 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |