Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119262.6 | NP_194841.2 | 22 - 23 | 1969 | 101 | |
NM_202918.2 | NP_974647.1 | 21 - 22 | 2004 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |