Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRE11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345081.1 | NP_001330550.1 | 4 - 5 | 533 | 137 | |
NM_124806.2 | NP_200237.1 | 6 - 7 | 719 | 137 | ||
GenBank mRNA | AJ243822.1 | - | 6 - 7 | 688 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |