Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124811.5 | NP_568807.1 | 1 - 2 | 261 | 71 | |
GenBank mRNA | AK226402.1 | - | 1 - 2 | 69 | 71 | |
AY099716.1 | - | 1 - 2 | 207 | 71 | ||
BT000287.1 | - | 1 - 2 | 57 | 71 | ||
FJ794601.1 | - | 1 - 2 | 159 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Arabidopsis thaliana Probe spans exons |