Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036999.1 | NP_001032076.1 | 8 - 9 | 746 | 94 | |
NM_001345117.1 | NP_001330104.1 | 6 - 7 | 561 | 94 | ||
NM_124895.3 | NP_200324.1 | 8 - 9 | 879 | 94 | ||
GenBank mRNA | AF124159.1 | - | 8 - 9 | 796 | 94 | |
AK317295.1 | - | 8 - 9 | 743 | 94 | ||
BT029156.1 | - | 8 - 9 | 769 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |