Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IAMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085287.2 | NP_001078756.1 | 3 - 4 | 879 | 56 | |
NM_124907.5 | NP_200336.1 | 3 - 4 | 927 | 56 | ||
GenBank mRNA | AK175586.1 | - | 3 - 4 | 871 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Arabidopsis thaliana Probe spans exons |