Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124958.3 | NP_200387.1 | 9 - 10 | 826 | 94 | |
GenBank mRNA | AF283757.1 | - | 9 - 10 | 786 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |