Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NMNAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203622.1 | NP_001190551.1 | 8 - 9 | 805 | 154 | |
NM_001345151.1 | NP_001329970.1 | 7 - 8 | 896 | 154 | ||
NM_124963.5 | NP_200392.3 | 8 - 9 | 785 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |