Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345176.1 | NP_001332655.1 | 8 - 9 | 1029 | 88 | |
NM_124981.4 | NP_200410.1 | 8 - 9 | 1029 | 88 | ||
GenBank mRNA | AF076252.1 | - | 7 - 8 | 567 | 88 | |
AY052304.1 | - | 7 - 8 | 612 | 88 | ||
AY139808.1 | - | 6 - 7 | 503 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |