Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124990.2 | NP_200419.1 | 1 | 936 | 86 | |
GenBank mRNA | BT030013.1 | - | 1 | 859 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |