Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036636.2 | NP_001031713.1 | 4 - 5 | 598 | 69 | |
NM_001341732.1 | NP_001329739.1 | 4 - 5 | 577 | 69 | ||
NM_001341733.1 | NP_001329738.1 | 4 - 5 | 549 | 69 | ||
NM_001341734.1 | NP_001329741.1 | 4 - 5 | 549 | 69 | ||
NM_001341735.1 | NP_001329740.1 | 3 - 4 | 707 | 69 | ||
NM_001341736.1 | NP_001329746.1 | 2 - 3 | 967 | 69 | ||
NM_001341737.1 | NP_001329743.1 | 2 - 3 | 952 | 69 | ||
NM_001341738.1 | NP_001329742.1 | 3 - 4 | 873 | 69 | ||
NM_001341739.1 | NP_001329745.1 | 4 - 5 | 577 | 69 | ||
NM_001341740.1 | NP_001329744.1 | 4 - 5 | 577 | 69 | ||
NM_001341741.1 | NP_001329748.1 | 4 - 5 | 577 | 69 | ||
NM_001341742.1 | NP_001329747.1 | 4 - 5 | 562 | 69 | ||
NM_179112.3 | NP_849443.2 | 4 - 5 | 562 | 69 | ||
GenBank mRNA | BT010765.1 | - | 4 - 5 | 526 | 69 | |
BT010987.1 | - | 4 - 5 | 434 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |