Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342113.1 | NP_001320109.1 | 2 - 3 | 948 | 139 | |
NM_001342114.1 | NP_001329161.1 | 2 - 3 | 935 | 139 | ||
NM_119327.3 | NP_194906.1 | 2 - 3 | 924 | 139 | ||
GenBank mRNA | AB047399.1 | - | 2 - 3 | 805 | 139 | |
AF241797.1 | - | 2 - 3 | 860 | 139 | ||
AY092965.1 | - | 2 - 3 | 1001 | 139 | ||
BT008890.1 | - | 2 - 3 | 701 | 139 | ||
KJ138730.1 | - | 2 - 3 | 701 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |