Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100030.4 | NP_171655.1 | 2 - 3 | 456 | 84 | |
NM_179241.1 | NP_849572.1 | 1 - 2 | 195 | 84 | ||
GenBank mRNA | AF334719.1 | - | 2 - 3 | 300 | 84 | |
AY052207.1 | - | 1 - 2 | 195 | 84 | ||
M95595.1 | - | 2 - 3 | 300 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |