Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UVH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100201.3 | NP_171818.1 | 7 - 8 | 1810 | 112 | |
NM_179253.1 | NP_849584.1 | 6 - 7 | 1873 | 112 | ||
GenBank mRNA | AY062471.1 | - | 7 - 8 | 1810 | 112 | |
AY090788.1 | - | 6 - 7 | 1873 | 112 | ||
AY090915.1 | - | 6 - 7 | 1673 | 112 | ||
BT010344.1 | - | 6 - 7 | 1673 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |