Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ML5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332851.1 | NP_001320501.1 | 5 - 6 | 1361 | 116 | |
NM_102680.2 | NP_174233.2 | 5 - 6 | 969 | 116 | ||
NM_179396.3 | NP_849727.1 | 5 - 6 | 1118 | 116 | ||
GenBank mRNA | AY062613.1 | - | 5 - 6 | 1005 | 116 | |
AY070368.1 | - | 5 - 6 | 1008 | 116 | ||
AY091327.1 | - | 4 - 5 | 845 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |