Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112177.3 | NP_566452.1 | 2 - 3 | 515 | 118 | |
GenBank mRNA | AF424628.1 | - | 2 - 3 | 200 | 118 | |
AY097343.1 | - | 1 - 2 | 51 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |