Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112191.3 | NP_187954.1 | 6 - 7 | 766 | 77 | |
GenBank mRNA | AF145452.1 | - | 6 - 7 | 703 | 77 | |
AK229269.1 | - | 6 - 7 | 764 | 77 | ||
BT024889.1 | - | 6 - 7 | 625 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |