Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OVA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112195.3 | NP_187958.1 | 8 - 9 | 1086 | 71 | |
GenBank mRNA | BT002317.1 | - | 8 - 9 | 1038 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |