Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112210.3 | NP_187973.1 | 6 - 7 | 699 | 112 | |
GenBank mRNA | BT003947.1 | - | 6 - 7 | 583 | 112 | |
BT005042.1 | - | 5 - 6 | 493 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |