Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112225.5 | NP_187988.1 | 6 - 7 | 925 | 61 | |
GenBank mRNA | AJ270297.1 | - | 6 - 7 | 903 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |