Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAP1;4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338075.1 | NP_001319544.1 | 8 - 9 | 1015 | 98 | |
NM_112230.3 | NP_187993.2 | 7 - 8 | 896 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |