Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112241.4 | NP_188003.1 | 7 - 8 | 761 | 80 | |
NM_202579.1 | NP_974308.1 | 7 - 8 | 670 | 80 | ||
GenBank mRNA | AY092988.1 | - | 7 - 8 | 605 | 80 | |
AY128801.1 | - | 6 - 7 | 515 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |